siRNA Details
| Gene Symbol: MLLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005934.3 | NP_005925.2 | 3 | 420 | ||
| XM_011528021.1 | XP_011526323.1 | 3 | 394 | |||
| XM_011528022.1 | XP_011526324.1 | 3 | 277 | |||
| XM_011528023.1 | XP_011526325.1 | 3 | 394 | |||
| XM_017026819.1 | XP_016882308.1 | 3 | 976 | |||
| GenBank mRNA | AF373587.1 | - | 227 | |||
| D14539.1 | - | 3 | 420 | |||
| L04285.1 | - | 2 | 256 | |||

MLLT1
Genomic Map