siRNA Details
| Gene Symbol: NR3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000901.4 | NP_000892.2 | 2 | 685 | ||
| NM_001166104.1 | NP_001159576.1 | 2 | 685 | |||
| XM_011531975.1 | XP_011530277.1 | 2 | 685 | |||
| XM_011531976.2 | XP_011530278.1 | 2 | 982 | |||
| XM_011531977.2 | XP_011530279.1 | 2 | 412 | |||
| XM_011531978.2 | XP_011530280.1 | 2 | 685 | |||
| XM_017008217.1 | XP_016863706.1 | 2 | 982 | |||
| GenBank mRNA | AJ315514.1 | - | 2 | 536 | ||
| AK304318.1 | - | 2 | 687 | |||
| BC111758.1 | - | 2 | 389 | 40027353 | ||
| M16801.1 | - | 2 | 538 | |||

NR3C2
Genomic Map