siRNA Details
| Gene Symbol: TMEM198B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036476.1 | 3 | 1376 | |||
| NR_036478.1 | 3 | 1376 | ||||
| NR_036479.1 | 3 | 1376 | ||||
| GenBank mRNA | AK057179.1 | - | 2 | 204 | ||
| AK095340.1 | - | 3 | 1239 | |||
| BC072431.1 | - | 2 | 264 | |||

TMEM198B
Genomic Map