siRNA Details
| Gene Symbol: ALG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004127.2 | NP_001004127.2 | 3 | 347 | ||
| GenBank mRNA | AK025456.1 | - | 3 | 320 | ||
| AK128503.1 | - | 2 | 3252 | |||
| AK296747.1 | - | 3 | 337 | |||
| BC111022.1 | - | 3 | 320 | |||
| BC142998.1 | - | 3 | 346 | 8860455 | ||
| DC366525.1 | - | 3 | 348 | |||

ALG11
Genomic Map