siRNA Details
| Gene Symbol: MZT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001071775.2 | NP_001065243.1 | 2 | 390 | ||
| GenBank mRNA | AK025522.1 | - | 2 | 263 | ||
| BC062361.1 | - | 2 | 223 | |||
| BC080523.1 | - | 2 | 239 | |||
| BC125183.1 | - | 2 | 393 | 40122127 | ||

MZT1
Genomic Map