siRNA Details
| Gene Symbol: UNC13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080534.1 | NP_001074003.1 | 22 | 5326 | ||
| XM_005254394.4 | XP_005254451.1 | 23 | 6277 | |||
| XM_017022220.1 | XP_016877709.1 | 27 | 6197 | |||
| XM_017022221.1 | XP_016877710.1 | 26 | 6077 | |||
| XM_017022222.1 | XP_016877711.1 | 25 | 5975 | |||
| XM_017022223.1 | XP_016877712.1 | 23 | 11503 | |||
| XM_017022224.1 | XP_016877713.1 | 22 | 6203 | |||
| XM_017022225.1 | XP_016877714.1 | 20 | 3628 | |||
| GenBank mRNA | AK091491.1 | - | 19 | 2302 | ||
| AK293996.1 | - | 12 | 1243 | |||
| AL834407.1 | - | 6 | 601 | |||

UNC13C
Genomic Map