siRNA Details
| Gene Symbol: GPR179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004334.3 | NP_001004334.3 | 11 | 3598 | ||
| GenBank mRNA | AY584611.1 | - | 11 | 3598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR179| Gene Symbol: GPR179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004334.3 | NP_001004334.3 | 11 | 3598 | ||
| GenBank mRNA | AY584611.1 | - | 11 | 3598 | ||