siRNA Details
| Gene Symbol: ODF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014440.3 | NP_001014440.2 | 5 | 653 | ||
| GenBank mRNA | BC127934.1 | - | 1 | 1154 | ||
| BC127935.1 | - | 4 | 482 | |||
| BG149596.1 | - | 3 | 386 | |||
| BM973820.1 | - | 3 | 315 | |||
| S72487.1 | - | 2 | 726 | |||

ODF3B
Genomic Map