siRNA Details
| Gene Symbol: ODF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014440.3 | NP_001014440.2 | 6, 7 | 811 | ||
| GenBank mRNA | BC127935.1 | - | 5, 6 | 640 | ||
| BM973820.1 | - | 1, 2 | 157 | |||
| S72487.1 | - | 3, 4 | 860 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ODF3B| Gene Symbol: ODF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014440.3 | NP_001014440.2 | 6, 7 | 811 | ||
| GenBank mRNA | BC127935.1 | - | 5, 6 | 640 | ||
| BM973820.1 | - | 1, 2 | 157 | |||
| S72487.1 | - | 3, 4 | 860 | |||