siRNA Details
| Gene Symbol: SMIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001124767.1 | NP_001118239.1 | 1 | 224 | ||
| XM_011533729.2 | XP_011532031.1 | 1 | 340 | |||
| GenBank mRNA | AK095910.1 | - | 1 | 224 | ||
| BC017996.1 | - | 1 | 224 | |||
| BG031013.1 | - | 1 | 214 | |||
| BM674004.1 | - | 2 | 240 | |||

SMIM4
Genomic Map