siRNA Details
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543306.2 | XP_011541608.1 | 1 | 247 | ||
XM_011543307.2 | XP_011541609.1 | 1 | 688 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543306.2 | XP_011541608.1 | 1 | 247 | ||
XM_011543307.2 | XP_011541609.1 | 1 | 688 |