siRNA Details
Gene Symbol: C5orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045116.1 | 2, 3 | 388 | |||
GenBank mRNA | AK096941.1 | - | 2, 3 | 180 | ||
AK128882.1 | - | 2, 3 | 177 | |||
AK309191.1 | - | 2, 3 | 180 | |||
BC130299.1 | - | 1, 2 | 78 | 40146330 | ||
BC130656.1 | - | 1, 2 | 78 | 40146687 | ||
BM998490.1 | - | 2, 3 | 430 | |||
DC350817.1 | - | 2, 3 | 214 | |||
DN991436.1 | - | 2, 3 | 396 |
