siRNA Details
Gene Symbol: ZNF716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159279.1 | NP_001152751.1 | 4 | 1199 | ||
GenBank mRNA | AK131575.1 | - | 4 | 1199 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159279.1 | NP_001152751.1 | 4 | 1199 | ||
GenBank mRNA | AK131575.1 | - | 4 | 1199 |