siRNA Details
| Gene Symbol: ANKRD18B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011517881.2 | XP_011516183.1 | 21 | 4956 | ||
| XM_011517883.2 | XP_011516185.1 | 20 | 4902 | |||
| XM_011517885.2 | XP_011516187.1 | 15 | 4047 | |||
| XM_017014728.1 | XP_016870217.1 | 15 | 4044 | |||

ANKRD18B
Genomic Map