siRNA Details
| Gene Symbol: CDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029954.2 | NP_001025125.2 | 2 | 224 | ||
| XM_011519488.2 | XP_011517790.1 | 2 | 313 | |||
| GenBank mRNA | AV657190.1 | - | 2 | 320 | ||
| BC037872.1 | - | 4 | 389 | |||
| BC133042.1 | - | 2 | 211 | 40147064 | ||
| BC133044.1 | - | 2 | 211 | 40147066 | ||
| BC144619.1 | - | 2 | 211 | |||
| CN311321.1 | - | 2 | 200 | |||

CDNF
Genomic Map