siRNA Details
| Gene Symbol: TSPAN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080509.2 | NP_001073978.1 | 3, 4 | 379 | ||
| XM_011520679.2 | XP_011518981.1 | 3, 4 | 641 | |||
| XM_011520680.2 | XP_011518982.1 | 3, 4 | 619 | |||
| GenBank mRNA | AK293947.1 | - | 3, 4 | 326 | ||
| AK300920.1 | - | 2, 3 | 284 | |||
| BC127660.1 | - | 1, 2 | 145 | |||
| BC127661.1 | - | 1, 2 | 145 | |||
| BC146917.1 | - | 3, 4 | 325 | 9056845 | ||
| BC146922.1 | - | 3, 4 | 325 | 9056850 | ||

TSPAN11
Genomic Map