siRNA Details
Gene Symbol: OR4Q3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172194.1 | NP_751944.1 | 1 | 900 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR4Q3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172194.1 | NP_751944.1 | 1 | 900 |