siRNA Details
| Gene Symbol: RFPL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013734.2 | NP_001013756.2 | 3 | 484 | ||
| XM_017010897.1 | XP_016866386.1 | 4 | 626 | |||
| XM_017010898.1 | XP_016866387.1 | 4 | 801 | |||
| GenBank mRNA | AK122906.1 | - | 3 | 484 | ||
| BC132796.1 | - | 2 | 315 | 40146818 | ||
| BC132798.1 | - | 2 | 315 | 40146820 | ||

RFPL4B
Genomic Map