siRNA Details
| Gene Symbol: LMOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207163.1 | NP_997046.1 | 2 | 949 | ||
| GenBank mRNA | AK299389.1 | - | 2 | 890 | ||
| AK300698.1 | - | 3 | 800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LMOD2| Gene Symbol: LMOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207163.1 | NP_997046.1 | 2 | 949 | ||
| GenBank mRNA | AK299389.1 | - | 2 | 890 | ||
| AK300698.1 | - | 3 | 800 | |||