siRNA Details
| Gene Symbol: LCE6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128600.1 | NP_001122072.1 | 2 | 307 | ||
| XM_017001327.1 | XP_016856816.1 | 2 | 228 | |||
| GenBank mRNA | BG210316.1 | - | 1 | 154 | ||
| DQ991251.1 | - | 2 | 307 | |||

LCE6A
Genomic Map