siRNA Details
| Gene Symbol: GADD45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015675.3 | NP_056490.2 | 4 | 653 | ||
| XM_017026822.1 | XP_016882311.1 | 3 | 887 | |||
| GenBank mRNA | AF078077.1 | - | 4 | 519 | ||
| AF087853.1 | - | 4 | 640 | |||
| AF090950.1 | - | 4 | 451 | |||
| AK129595.1 | - | 1 | 676 | |||
| AK293087.1 | - | 4 | 651 | |||
| BC024654.3 | - | 4 | 559 | |||
| BC113466.1 | - | 4 | 510 | 8322518 | ||
| BC117469.1 | - | 4 | 510 | 40126020 | ||

GADD45B
Genomic Map