siRNA Details
| Gene Symbol: MYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000432.3 | NP_000423.2 | 3 | 206 | ||
| GenBank mRNA | AF020768.1 | - | 2 | 140 | ||
| AK312013.1 | - | 3 | 183 | |||
| BC015821.1 | - | 3 | 188 | 4245364 | ||
| BC031006.1 | - | 3 | 183 | 4292208 | ||
| BC031008.1 | - | 3 | 185 | 4297454 | ||
| CR456962.1 | - | 2 | 135 | |||
| CR541957.1 | - | 2 | 135 | |||
| M22815.1 | - | 3 | 157 | |||
| S69022.1 | - | 3 | 212 | |||
| X14332.1 | - | 2 | 135 | |||
| X57542.1 | - | 3 | 157 | |||
| X66141.1 | - | 3 | 165 | |||

MYL2
Genomic Map