siRNA Details
| Gene Symbol: MYO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012334.2 | NP_036466.2 | 15 | 2015 | ||
| XM_006714475.2 | XP_006714538.1 | 15 | 1791 | |||
| GenBank mRNA | AB018342.2 | - | 15 | 2002 | ||
| AF132021.1 | - | 15 | 1627 | |||
| AF234532.1 | - | 15 | 1769 | |||
| AF247457.2 | - | 15 | 1700 | |||
| AK024719.1 | - | 6 | 626 | |||
| AK125246.1 | - | 15 | 1999 | |||
| AL832428.1 | - | 5 | 472 | |||
| BC014956.1 | - | 15 | 2011 | |||
| BC137168.1 | - | 15 | 1724 | 9021165 | ||
| BC150285.1 | - | 15 | 2002 | 8860110 | ||

MYO10
Genomic Map