siRNA Details
| Gene Symbol: NDUFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004541.3 | NP_004532.1 | 2, 3 | 339 | ||
| GenBank mRNA | AB451304.1 | - | 2, 3 | 198 | ||
| BC000266.1 | - | 2, 3 | 236 | 3352028 | ||
| BQ680674.1 | - | 2, 3 | 240 | |||
| CR456889.1 | - | 2, 3 | 198 | |||
| U54993.1 | - | 2, 3 | 231 | |||
| X81900.1 | - | 2, 3 | 215 | |||

NDUFA1
Genomic Map