siRNA Details
| Gene Symbol: NDUFS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002496.3 | NP_002487.1 | 5 | 452 | ||
| XM_005274013.1 | XP_005274070.1 | 5 | 549 | |||
| XM_005274014.2 | XP_005274071.1 | 5 | 576 | |||
| XM_011545053.2 | XP_011543355.1 | 5 | 487 | |||
| GenBank mRNA | AK002110.1 | - | 3 | 1040 | ||
| AK223114.1 | - | 5 | 442 | |||
| AK302449.1 | - | 5 | 452 | |||
| AK314546.1 | - | 5 | 377 | |||
| BC119754.2 | - | 5 | 381 | 40119290 | ||
| DB498707.1 | - | 5 | 438 | |||
| U65579.1 | - | 5 | 375 | |||

NDUFS8
Genomic Map