siRNA Details
| Gene Symbol: DRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004147.3 | NP_004138.1 | 2 | 212 | ||
| GenBank mRNA | AF078103.1 | - | 2 | 142 | ||
| AJ005940.1 | - | 2 | 146 | |||
| AK315659.1 | - | 2 | 170 | |||
| BC019285.1 | - | 2 | 151 | 2823263 | ||
| BC020803.1 | - | 2 | 146 | 4250998 | ||
| BT007237.1 | - | 2 | 81 | |||
| CR456488.1 | - | 2 | 123 | |||
| CR542059.1 | - | 2 | 81 | |||

DRG1
Genomic Map