siRNA Details
| Gene Symbol: NNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006169.2 | NP_006160.1 | 1 | 858 | ||
| GenBank mRNA | AK026845.1 | - | 1 | 234 | ||
| BC000234.1 | - | 3 | 378 | 3350358 | ||
| CR541907.1 | - | 1 | 114 | |||
| U08021.1 | - | 1 | 231 | |||

NNMT
Genomic Map