siRNA Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083592.1 | NP_001077061.1 | 6 | 1274 | ||
| NM_005012.3 | NP_005003.2 | 6 | 1274 | |||
| XM_011541526.1 | XP_011539828.1 | 6 | 873 | |||
| XM_017001376.1 | XP_016856865.1 | 5 | 1174 | |||
| XM_017001377.1 | XP_016856866.1 | 8 | 3817 | |||
| GenBank mRNA | AK308088.1 | - | 6 | 1280 | ||
| AK309189.1 | - | 3 | 516 | |||
| BC080541.1 | - | 6 | 1274 | |||
| BC128386.1 | - | 7 | 884 | |||
| M97675.1 | - | 6 | 1250 | |||

ROR1
Genomic Map