siRNA Details
| Gene Symbol: C8orf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007176.4 | NP_001007177.1 | 2 | 214 | ||
| NM_001256596.1 | NP_001243525.1 | 2 | 106 | |||
| NM_001256597.1 | NP_001243526.1 | 2 | 214 | |||
| NM_001256598.1 | NP_001243527.1 | 2 | 214 | |||
| GenBank mRNA | AJ276240.1 | - | 2 | 201 | ||
| BC017981.1 | - | 2 | 207 | 4245342 | ||
| BG202040.1 | - | 2 | 132 | |||
| BX500169.1 | - | 2 | 223 | |||
| DA820911.1 | - | 2 | 215 | |||
| DA900243.1 | - | 2 | 214 | |||

C8orf22
Genomic Map