siRNA Details
| Gene Symbol: CISD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008388.4 | NP_001008389.1 | 1 | 187 | ||
| GenBank mRNA | AK292134.1 | - | 1 | 124 | ||
| BC032300.1 | - | 1 | 124 | 5105935 | ||
| BX537971.1 | - | 1 | 125 | |||
| DN993861.1 | - | 1 | 192 | |||

CISD2
Genomic Map