siRNA Details
Gene Symbol: RNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256758.1 | NP_001243687.1 | 5 | 511 | ||
NM_152470.2 | NP_689683.2 | 7 | 935 | |||
XM_011526016.2 | XP_011524318.1 | 6 | 839 | |||
GenBank mRNA | AK122819.1 | - | 5 | 511 | ||
AK127467.1 | - | 7 | 935 |
