siRNA Details
| Gene Symbol: WEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105558.1 | NP_001099028.1 | 10 | 1849 | ||
| GenBank mRNA | AK131218.1 | - | 10 | 1849 | ||
| BG402079.1 | - | 2 | 199 | |||
| Gene Symbol: WEE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015392.1 | 4 | 393 | |||
| GenBank mRNA | AK098171.1 | - | 4 | 393 | ||
| BC101359.2 | - | 4 | 647 | |||

WEE2
Genomic Map