siRNA Details
Gene Symbol: OMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005014.2 | NP_005005.1 | 2 | 1246 | ||
GenBank mRNA | AB000114.1 | - | 2 | 977 | ||
AK225922.1 | - | 2 | 1091 | |||
AK313055.1 | - | 2 | 1085 | |||
AY358872.1 | - | 2 | 1144 | |||
BC046356.1 | - | 2 | 1148 | 5239983 |
