siRNA Details
| Gene Symbol: C18orf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035005.3 | NP_001030177.1 | 2 | 326 | ||
| NM_001199346.1 | NP_001186275.1 | 2 | 162 | |||
| GenBank mRNA | AB097012.1 | - | 2 | 246 | ||
| AK027111.1 | - | 2 | 122 | |||
| AK291882.1 | - | 2 | 133 | |||
| BC022357.1 | - | 2 | 131 | 4297017 | ||
| BC093004.1 | - | 2 | 283 | 5273608 | ||
| Gene Symbol: RPL17-C18orf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199355.1 | NP_001186284.1 | 6 | 614 | ||
| NM_001199356.1 | NP_001186285.1 | 6 | 681 | |||
| GenBank mRNA | BI461805.1 | - | 6 | 601 | ||
| BX393840.2 | - | 6 | 616 | |||

C18orf32
Genomic Map