siRNA Details
| Gene Symbol: CD207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015717.4 | NP_056532.4 | 2, 3 | 225 | ||
| XM_011532874.1 | XP_011531176.1 | 3, 4 | 413 | |||
| XM_011532875.2 | XP_011531177.1 | 4, 5 | 430 | |||
| XM_011532876.2 | XP_011531178.1 | 4, 5 | 429 | |||
| GenBank mRNA | AJ242859.1 | - | 2, 3 | 225 | ||
| AK314927.1 | - | 2, 3 | 219 | |||
| BC022278.1 | - | 2, 3 | 218 | 4692155 | ||

CD207
Genomic Map