siRNA Details
| Gene Symbol: DEF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022047.3 | NP_071330.3 | 6 | 869 | ||
| GenBank mRNA | AJ276095.1 | - | 6 | 830 | ||
| AK093613.1 | - | 5 | 581 | |||
| AK314756.1 | - | 6 | 844 | |||
| AY241694.1 | - | 6 | 869 | |||
| BC017504.1 | - | 6 | 835 | |||

DEF6
Genomic Map