siRNA Details
| Gene Symbol: DEF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022047.3 | NP_071330.3 | 6 | 910 | ||
| GenBank mRNA | AJ276095.1 | - | 6 | 871 | ||
| AK093613.1 | - | 5 | 622 | |||
| AK314756.1 | - | 6 | 885 | |||
| AY241694.1 | - | 6 | 910 | |||
| BC017504.1 | - | 6 | 876 | |||

DEF6
Genomic Map