siRNA Details
| Gene Symbol: CYHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129888.1 | NP_001123360.1 | 3 | 441 | ||
| NM_032687.3 | NP_116076.1 | 3 | 477 | |||
| NM_138496.1 | NP_612505.1 | 2 | 302 | |||
| XM_006716560.3 | XP_006716623.1 | 3 | 694 | |||
| XM_006716561.3 | XP_006716624.1 | 3 | 646 | |||
| GenBank mRNA | BC056412.1 | - | 2 | 755 | ||

CYHR1
Genomic Map