siRNA Details
| Gene Symbol: SLC45A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080397.2 | NP_001073866.2 | 5 | 1216 | ||
| XM_011541530.1 | XP_011539832.1 | 5 | 1193 | |||
| XM_011541531.1 | XP_011539833.1 | 4 | 1100 | |||
| GenBank mRNA | AF118274.1 | - | 1 | 445 | ||

SLC45A1
Genomic Map