siRNA Details
Gene Symbol: ZNF593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015871.4 | NP_056955.2 | 2 | 322 | ||
XM_017001398.1 | XP_016856887.1 | 2 | 691 | |||
GenBank mRNA | AI083892.1 | - | 2 | 328 | ||
AK311926.1 | - | 2 | 322 | |||
BC002580.2 | - | 2 | 300 | 3143091 | ||
BC019267.1 | - | 2 | 295 | 2823658 | ||
D45213.1 | - | 2 | 231 | |||
DA124660.1 | - | 2 | 322 |
