siRNA Details
| Gene Symbol: SIDT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040455.1 | NP_001035545.1 | 5 | 644 | ||
| GenBank mRNA | AF151799.1 | - | 5 | 686 | ||
| AK292501.1 | - | 5 | 1124 | |||
| AK292974.1 | - | 5 | 635 | |||
| AK303023.1 | - | 5 | 600 | |||
| AY358442.1 | - | 5 | 653 | |||
| BC114522.1 | - | 5 | 673 | 40035836 | ||
| KC109742.1 | - | 5 | 600 | |||

SIDT2
Genomic Map