siRNA Details
| Gene Symbol: ANKRD39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016466.5 | NP_057550.3 | 2 | 169 | ||
| GenBank mRNA | AF151034.1 | - | 2 | 149 | ||
| AK312193.1 | - | 2 | 166 | |||
| BC031303.2 | - | 2 | 161 | 5284947 | ||
| BX091421.1 | - | 2 | 169 | |||
| DA769410.1 | - | 2 | 166 | |||

ANKRD39
Genomic Map