siRNA Details
| Gene Symbol: RASL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307930.1 | NP_001294859.1 | 2 | 397 | ||
| NM_016563.3 | NP_057647.1 | 3 | 454 | |||
| XM_005254434.4 | XP_005254491.1 | 3 | 264 | |||
| XM_011521660.2 | XP_011519962.1 | 4 | 267 | |||
| XM_017022296.1 | XP_016877785.1 | 3 | 264 | |||
| GenBank mRNA | AF233588.1 | - | 3 | 293 | ||
| AK296261.1 | - | 2 | 207 | |||
| AK300536.1 | - | 3 | 237 | |||
| AK314914.1 | - | 3 | 262 | |||
| BC053734.1 | - | 3 | 302 | 5588236 | ||
| BI489884.1 | - | 3 | 454 | |||

RASL12
Genomic Map