siRNA Details
| Gene Symbol: CEND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016564.3 | NP_057648.2 | 2 | 391 | ||
| GenBank mRNA | AF235030.1 | - | 1 | 301 | ||
| AK074547.1 | - | 2 | 388 | |||
| AK312993.1 | - | 2 | 388 | |||
| BC034732.1 | - | 2 | 378 | 5179740 | ||
| DA066200.1 | - | 2 | 391 | |||

CEND1
Genomic Map