siRNA Details
| Gene Symbol: ERGIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016570.2 | NP_057654.2 | 10 | 809 | ||
| GenBank mRNA | AF183410.1 | - | 10 | 763 | ||
| AF216751.1 | - | 10 | 767 | |||
| AF302767.1 | - | 10 | 762 | |||
| AK074520.1 | - | 10 | 784 | |||
| AK222799.1 | - | 10 | 749 | |||
| AL834128.2 | - | 10 | 750 | |||
| BC000887.1 | - | 10 | 762 | 3462408 | ||
| BC107794.1 | - | 10 | 766 | 30319807 | ||

ERGIC2
Genomic Map