siRNA Details
Gene Symbol: TEX264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129884.2 | NP_001123356.1 | 3 | 433 | ||
NM_001243725.1 | NP_001230654.1 | 2 | 541 | |||
NM_001243726.1 | NP_001230655.1 | 2 | 325 | |||
NM_001243727.2 | NP_001230656.1 | 2 | 381 | |||
NM_001278195.1 | NP_001265124.1 | 3 | 473 | |||
NM_015926.5 | NP_057010.1 | 2 | 381 | |||
XM_006713195.3 | XP_006713258.1 | 2 | 321 | |||
XM_006713197.3 | XP_006713260.1 | 2 | 288 | |||
XM_006713198.2 | XP_006713261.1 | 1 | 267 | |||
XM_011533805.2 | XP_011532107.1 | 2 | 1036 | |||
XM_017006573.1 | XP_016862062.1 | 2 | 541 | |||
XM_017006574.1 | XP_016862063.1 | 1 | 267 | |||
GenBank mRNA | AF072733.1 | - | 1 | 279 | ||
AK023997.1 | - | 2 | 324 | |||
AK222938.1 | - | 2 | 293 | |||
AK222949.1 | - | 2 | 293 | |||
AY358972.1 | - | 1 | 280 | |||
BC001592.1 | - | 2 | 303 | |||
BC008742.2 | - | 2 | 541 | 3346733 | ||
BG326586.1 | - | 2 | 298 | |||
BI826172.1 | - | 3 | 393 | |||
BP252919.1 | - | 3 | 363 | |||
DA894396.1 | - | 2 | 350 | |||
HY149215.1 | - | 2 | 297 |
