siRNA Details
| Gene Symbol: PDE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003683.2 | NP_001003683.1 | 4 | 480 | ||
| NM_001258312.1 | NP_001245241.1 | 4 | 507 | |||
| NM_001258313.1 | NP_001245242.1 | 3 | 659 | |||
| NM_001258314.1 | NP_001245243.1 | 2 | 336 | |||
| NM_005019.4 | NP_005010.2 | 3 | 522 | |||
| XM_011511323.2 | XP_011509625.1 | 3 | 299 | |||
| XM_011511324.2 | XP_011509626.1 | 4 | 527 | |||
| XM_011511325.2 | XP_011509627.1 | 3 | 654 | |||
| XM_011511326.2 | XP_011509628.1 | 2 | 331 | |||
| XM_017004294.1 | XP_016859783.1 | 3 | 522 | |||
| XM_017004295.1 | XP_016859784.1 | 4 | 527 | |||
| XM_017004296.1 | XP_016859785.1 | 4 | 449 | |||
| XM_017004297.1 | XP_016859786.1 | 4 | 390 | |||
| XM_017004298.1 | XP_016859787.1 | 4 | 572 | |||
| XM_017004299.1 | XP_016859788.1 | 4 | 527 | |||
| XM_017004300.1 | XP_016859789.1 | 4 | 572 | |||
| XM_017004301.1 | XP_016859790.1 | 4 | 377 | |||
| XM_017004302.1 | XP_016859791.1 | 3 | 654 | |||
| GenBank mRNA | AB038228.1 | - | 2 | 213 | ||
| AJ401610.1 | - | 3 | 241 | |||
| AK294239.1 | - | 4 | 442 | |||
| AK295657.1 | - | 4 | 312 | |||
| AK301720.1 | - | 2 | 319 | |||
| AL110263.1 | - | 3 | 633 | |||
| BC022480.1 | - | 3 | 490 | 4814064 | ||
| DA128734.1 | - | 3 | 523 | |||
| DA295167.1 | - | 4 | 530 | |||
| DC396228.1 | - | 2 | 336 | |||
| U40370.1 | - | 4 | 373 | |||

PDE1A
Genomic Map