siRNA Details
| Gene Symbol: CRLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015986.3 | NP_057070.3 | 2 | 365 | ||
| XM_011524879.1 | XP_011523181.1 | 2 | 539 | |||
| GenBank mRNA | AF046059.1 | - | 2 | 278 | ||
| AF120151.1 | - | 1 | 731 | |||
| AK313315.1 | - | 2 | 354 | |||
| BC023567.2 | - | 2 | 287 | 4309046 | ||
| BU902521.1 | - | 2 | 406 | |||
| DQ298450.1 | - | 2 | 244 | |||

CRLF3
Genomic Map