siRNA Details
| Gene Symbol: NOL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317724.1 | NP_001304653.1 | 5 | 494 | ||
| NM_016167.4 | NP_057251.2 | 5 | 494 | |||
| GenBank mRNA | AF172066.3 | - | 5 | 494 | ||
| AK075301.1 | - | 5 | 477 | |||
| AL050299.1 | - | 4 | 194 | |||
| BC023517.2 | - | 5 | 469 | 4298095 | ||
| BC062683.1 | - | 5 | 490 | 6499123 | ||
| BU071935.1 | - | 5 | 247 | |||

NOL7
Genomic Map