siRNA Details
| Gene Symbol: NAT8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016347.2 | NP_057431.2 | 1 | 404 | ||
| NR_132338.1 | 1 | 404 | ||||
| GenBank mRNA | AF185571.1 | - | 1 | 404 | ||
| BC069564.1 | - | 1 | 404 | |||
| BC121101.1 | - | 1 | 408 | |||
| BC121102.1 | - | 1 | 408 | |||
| Gene Symbol: LOC100996461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK304146.1 | - | 1 | 505 | ||

NAT8B
Genomic Map